Learn More
Waters Corporation XSelect HSS C18 Column
The HSS C18 is a general-purpose C18 bonded phase column that provides superior peak shape for bases, increased retention, and exceptional column lifetimes at low pH.
£942.00 - £1252.50
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 100 Å |
Quantity | 1/Ea. |
USP Type | L1 |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
15638436
![]() |
Waters Corporation
186004768 |
150 mm | 4.6 mm | 3.5 μm |
£1149.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17122912
![]() |
Waters Corporation
186004767 |
100 mm | 4.6 mm | 3.5 μm |
£1074.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17132912
![]() |
Waters Corporation
186004770 |
250 mm | 4.6 mm | 3.5 μm |
£1252.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17172912
![]() |
Waters Corporation
186004775 |
250 mm | 4.6 mm | 5 μm |
£1218.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17133672
![]() |
Waters Corporation
186006384 |
150 mm | 2.1 mm | 3.5 μm |
£1149.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17153662
![]() |
Waters Corporation
186006376 |
50 mm | 1 mm | 3.5 μm |
£999.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17163662
![]() |
Waters Corporation
186006377 |
100 mm | 1 mm | 3.5 μm |
£1074.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17173662
![]() |
Waters Corporation
186006378 |
150 mm | 1 mm | 3.5 μm |
£1149.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17193662
![]() |
Waters Corporation
186006380 |
30 mm | 2.1 mm | 3.5 μm |
£970.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17103672
![]() |
Waters Corporation
186006381 |
50 mm | 2.1 mm | 3.5 μm |
£999.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17113672
![]() |
Waters Corporation
186006382 |
75 mm | 2.1 mm | 3.5 μm |
£1017.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17123672
![]() |
Waters Corporation
186006383 |
100 mm | 2.1 mm | 3.5 μm |
£1074.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17143672
![]() |
Waters Corporation
186006387 |
75 mm | 4.6 mm | 3.5 μm |
£1017.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17153672
![]() |
Waters Corporation
186006390 |
30 mm | 2.1 mm | 5 μm |
£942.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17163672
![]() |
Waters Corporation
186006391 |
50 mm | 2.1 mm | 5 μm |
£964.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17173672
![]() |
Waters Corporation
186006392 |
100 mm | 2.1 mm | 5 μm |
£1039.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17183672
![]() |
Waters Corporation
186006393 |
150 mm | 2.1 mm | 5 μm |
£1108.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17193672
![]() |
Waters Corporation
186006396 |
50 mm | 3 mm | 5 μm |
£964.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17103682
![]() |
Waters Corporation
186006397 |
100 mm | 3 mm | 5 μm |
£1039.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17113682
![]() |
Waters Corporation
186006398 |
150 mm | 3 mm | 5 μm |
£1108.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17123682
![]() |
Waters Corporation
186006399 |
250 mm | 3 mm | 5 μm |
£1218.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17143682
![]() |
Waters Corporation
186006401 |
30 mm | 4.6 mm | 5 μm |
£942.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17153682
![]() |
Waters Corporation
186006402 |
75 mm | 4.6 mm | 5 μm |
£999.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17163682
![]() |
Waters Corporation
186006403 |
100 mm | 4.6 mm | 5 μm |
£1039.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17182902
![]() |
Waters Corporation
186004762 |
100 mm | 3 mm | 3.5 μm |
£1074.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17192902
![]() |
Waters Corporation
186004763 |
150 mm | 3 mm | 3.5 μm |
£1149.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17102912
![]() |
Waters Corporation
186004765 |
30 mm | 3 mm | 3.5 μm |
£970.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17112912
![]() |
Waters Corporation
186004766 |
50 mm | 3 mm | 3.5 μm |
£999.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17142912
![]() |
Waters Corporation
186004771 |
30 mm | 4.6 mm | 3.5 μm |
£970.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17152912
![]() |
Waters Corporation
186004772 |
50 mm | 4.6 mm | 3.5 μm |
£999.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17162912
![]() |
Waters Corporation
186004773 |
150 mm | 4.6 mm | 5 μm |
£1108.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17152932
![]() |
Waters Corporation
186004852 |
50 mm | 4.6 mm | 5 μm |
£964.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17103232
![]() |
Waters Corporation
186005642 |
75 mm | 3 mm | 3.5 μm |
£1017.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- The HSS C18 packing is the most stable commercially available bonded phase and does not suffer from peak shape issues that plague older stationary phases that rely upon steric protection (and lack of endcapping) to produce stability at low pH.
- The HSS C18 HPLC columns are fully scalable and are completely transferable to UPLC separations.
Specifications
6000 psi (415 Bar) | |
100 Å | |
L1 | |
Column |
Spherical | |
1/Ea. | |
Yes |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.