Learn More
Waters Corporation XTerra Shield RP8 Prep Column
XTerra Preparative RP8 columns are hybrid-based, reversed-phase C8 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds.
£1264.50 - £17122.50
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 125 Å |
Quantity | 1/Ea. |
USP Type | L7 |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | |||
13134539
![]() |
Waters Corporation
186001155 |
50 mm | 7.8 mm | 5 μm |
£1264.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13174539
![]() |
Waters Corporation
186001159 |
100 mm | 7.8 mm | 5 μm |
£1585.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13114549
![]() |
Waters Corporation
186001163 |
150 mm | 7.8 mm | 10 μm |
£1689.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13144549
![]() |
Waters Corporation
186001167 |
30 mm | 7.8 mm | 10 μm |
£2424.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13144659
![]() |
Waters Corporation
186001478 |
150 mm | 7.8 mm | 5 μm |
£1896.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13107689
![]() |
Waters Corporation
186001937 |
100 mm | 19 mm | 5 μm |
£3481.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13147689
![]() |
Waters Corporation
186001941 |
50 mm | 30 mm | 5 μm |
£4297.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13187689
![]() |
Waters Corporation
186001945 |
100 mm | 30 mm | 5 μm |
£6778.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13260190
![]() |
Waters Corporation
186002221 |
50 mm | 50 mm | 5 μm |
£9180.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13200200
![]() |
Waters Corporation
186002225 |
100 mm | 50 mm | 5 μm |
£12523.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13290200
![]() |
Waters Corporation
186002258 |
150 mm | 19 mm | 10 μm |
£3964.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13230210
![]() |
Waters Corporation
186002262 |
250 mm | 19 mm | 10 μm |
£5032.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13270210
![]() |
Waters Corporation
186002266 |
30 mm | 19 mm | 10 μm |
£6033.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13210220
![]() |
Waters Corporation
186002270 |
150 mm | 30 mm | 10 μm |
£9088.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13250220
![]() |
Waters Corporation
186002274 |
250 mm | 30 mm | 10 μm |
£14247.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13290220
![]() |
Waters Corporation
186002278 |
30 mm | 30 mm | 10 μm |
£17122.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13230230
![]() |
Waters Corporation
186002282 |
50 mm | 50 mm | 10 μm |
£8215.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13230260
![]() |
Waters Corporation
186002382 |
150 mm | 19 mm | 5 μm |
£4171.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13210270
![]() |
Waters Corporation
186002390 |
75 mm | 30 mm | 5 μm |
£5526.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- These first generation columns provide superior pH stability compared to a silica-based column.
Specifications
6000 psi (415 Bar) | |
125 Å | |
L7 | |
Preparative Column |
Spherical | |
1/Ea. | |
Yes |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.