missing translation for 'onlineSavingsMsg'
Learn More

Xylene, Extra Pure, SLR, Fisher Chemical™

Product Code. 10784001 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic-coated Glass Bottle
Unit Size:
2.5 L
25 L
5 L
500mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10784001

Brand: Fisher Chemical X/0200/17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Xylene, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
MDL Number 77264

Specifications

Melting Point -34°C
Color Colorless
Boiling Point 136°C
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 8mbar at 20°C
Viscosity 0.6 mPaS at 20°C
Formula Weight 106.17g/mol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Xylene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Aromatic hydrocarbons, C8; Light Oil Redistillate, high boiling, Xylenes (o-, m-, p- isomers), Ethyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.