missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, ≥98.5%, ACS Reagent, xylenes plus ethylbenzene basis, Honeywell™ Riedel-de Haën™

Catalog Number 15692690
Click to view available options
Quantity:
500 mL
1 L
2.5 L
20 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
1330-20-7
MFCD00077264
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
1330-20-7
MFCD00077264

ACS Reagent,≥98.5% xylenes + ethylbenzene basis

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Melting Point -48°C
Density 0.86g/mL at 25°C
Boiling Point 137°C to 140°C
Quantity 2.5 L
Explosion Limit 0.07
Flash Point 25°C (77°F)
Packaging Glass Bottle
Physical Form Liquid
Linear Formula C6H4(CH3)2
Vapor Pressure 18mmHg (37.7°C)
UN Number UN1307
Formula Weight 106.17g/mol
Percent Purity ≥98.5% xylenes plus ethylbenzene basis
Grade ACS Reagent
Chemical Name or Material Xylenes
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'