missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenes, 98+%, for analysis, mixture of isomers , Thermo Scientific Chemicals
£25.50 - £76.30
Chemical Identifiers
CAS | 1330-20-7, 100-41-4 |
---|---|
Molecular Formula | C8H10 |
Synonym | Dimethylbenzene |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10338882
|
Thermo Scientific Acros
383940010 |
1 L |
£25.50
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15701819
|
Thermo Scientific Acros
383940050 |
5 L |
£76.30
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1330-20-7, 100-41-4 | |
C8H10 | |
Dimethylbenzene |
Specifications
-34.0°C | |
0.8650g/mL | |
23°C | |
0.865 | |
C6H4(CH3)2 | |
Dimethylbenzene | |
106.17 | |
Analytical |
1330-20-7, 100-41-4 | |
136.0°C to 140.0°C | |
98+% | |
C8H10 | |
0.6 mPa.s (20°C) | |
Solubility in water: 0.1g/L water (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf, methylene chloride, acetone and ether | |
98+% | |
Xylenes |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Xylenes
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only