missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenes, +98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -34.0°C |
Color | Colorless |
Density | 0.8650g/mL |
Boiling Point | 136.0°C to 140.0°C |
Quantity | 100 mL |
Flash Point | 23°C |
Packaging | AcroSeal™ Glass Bottle |
Physical Form | Liquid |
Assay Percent Range | 98+% |
Residue after Evaporation | 0.0003% max. |
Show More |
Safety and Handling
Product Identifier
- Xylenes
- Danger
- Acute toxicity Category 4
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'