missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenes, Histological Grade, Honeywell™
Histological Grade
£20.33 - £76.43
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15635760
|
Honeywell Research Chemicals
534056-500ML |
500 mL | Glass Bottle | N/A | |||||
15618420
|
Honeywell Research Chemicals
534056-4L |
4 L | Aluminum Bottle |
£76.43
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Commonly used for tissue sampling and processing
- Precise control of the manufacturing process ensures unsurpassed lot-to-lot consistency
- Available in a variety of packaging options (amber glass bottles, carbon steel flex-spout cans and carbon steel cans with NPT threads)
Chemical Identifiers
1330-20-7 | |
MFCD00077264 |
Specifications
1330-20-7 | |
0.86g/mL at 25°C | |
0.07 | |
Liquid | |
C6H4(CH3)2 | |
18mmHg (37.7°C) | |
106.17g/mol | |
Xylenes |
Colorless | |
140°C | |
25°C (77°F) | |
n20/D 1.497 | |
MFCD00077264 | |
UN1307 | |
Histological |
Safety and Handling
P210-P260-P280-P301 + P310-P305 + P351 + P338-P370 + P378
missing translation for 'dotInformation' : class 3 - PG 3 - Xylenes
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Xylenes
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- Aspiration toxicity Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H312 + H332-Harmful in contact with skin or if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title