missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, Honeywell

Puriss, for analysis (isomeric mixture)

£93.34 - £728.58

Chemical Identifiers

CAS 1330-20-7
MDL Number MFCD00077264
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15601560
View Documents Promotion Details
Honeywell Research Chemicals
16446-2.5L
2.5 L
£93.34
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15611560
View Documents
Honeywell Research Chemicals
16446-25L
2.5 L
£728.58
25 L
Please to purchase this item. Need a web account? Register with us today!
15681550
View Documents
Honeywell Research Chemicals
16446-10L
10 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications

Chemical Identifiers

1330-20-7
MFCD00077264

Specifications

1330-20-7
0.07
C6H4(CH3)2
18mmHg (37.7°C)
106.17g/mol
0.86g/mL at 25°C
25°C (77°F)
MFCD00077264
1307
Xylenes
Videos
Safety and Handling

Safety and Handling

P210-P260-P280-P301 + P310-P305 + P351 + P338-P370 + P378

H226-H304-H312 + H332-H315-H319-H335-H373

Danger

missing translation for 'einecsNumber' : 215-535-7

missing translation for 'rtecsNumber' : ZE2100000

Product Identifier
  • Xylenes

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312 + H332-Harmful in contact with skin or if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications