missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xylenol Orange, sodium salt

Product Code. 10421831
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3618-43-7
Molecular Formula C31H28N2Na4O13S

Specifications

Melting Point 210.0°C
Solubility Information Solubility in water: 510g/L (20°C).
Infrared Spectrum Authentic
Lambda Max 575 to 585nm (0.1N NaOH)
Formula Weight 760.59
Loss on Drying 10.0% (110°C, 1 hr) max.
Grade Pure
Packaging Glass bottle
Suitability (for Zinc titration) Passes test
UV Spectrometry Complies
Color Brown to Brown/Red
Quantity 10 g
Physical Form Crystalline Powder
Chemical Name or Material Xylenol Orange, sodium salt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.