missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange Tetrasodium Salt, Indicator for Metal Titration, Honeywell Fluka™

Product Code. p-7120905 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15644090

Brand: Honeywell-Fluka 338251G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Indicator for metal titration

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3618-43-7
MDL Number MFCD00044293

Specifications

Melting Point 195°C
UN Number NONH for all modes of transport
Formula Weight 760.58g/mol
Packaging Glass Bottle
Color Brown
Quantity 1 g
Physical Form Powder or Crystals
Chemical Name or Material Xylenol Orange tetrasodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.