missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Y-25130 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
A selective and potent 5-HT3 antagonist (Ki = 2.9 nM).
Specifications
Quantity | 50 mg |
Formula Weight | 386.28 |
Percent Purity | >99% |
Chemical Name or Material | Y-25130 hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'