Learn More
Thermo Scientific™ Y-PER™ Yeast Protein Extraction Reagent
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Y-PER Yeast Protein Extraction Reagent is the first commercially available yeast lysis reagent to use a mild detergent lysis procedure to efficiently release functionally active solubilized proteins.
Highlights:
More protein – extracts more than twice as much protein as glass bead methods
Less hassle – eliminates the difficulties associated with traditional glass bead lysis (e.g., clinging static-charged beads, protein/bead clumps and runaway beads)
Optmized for yeast – validated for use with Saccharomyces cerevisiae, Schizosaccharomyces pombe and Pichia pastoris
Versatile – effective for many different organisms, including Bacillus subtilus and both Gram-positive and Gram-negative bacteria; suitable for use in a diverse range of situations
Specifications
Specifications
| Quantity | 500 mL |
| Format | Liquid |
| Product Type | Yeast Protein Extraction Reagent |
| Volume (Metric) | 500 mL |
| Content And Storage | Store product at room temperature. |
| Product Line | Y-PER |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.