missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Y-PER™ Yeast Protein Extraction Reagent

Product Code. 11824121 Shop All Thermo Scientific Products
Click to view available options
Quantity:
200 mL
500 mL
Unit Size:
200mL
500mL
This item is not returnable. View return policy

Product Code. 11824121

Brand: Thermo Scientific™ 78991

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Release functionally active solubilized proteins with the first commercially available yeast lysis reagent to use a mild detergent lysis procedure.

Thermo Scientific™ Y-PER Yeast Protein Extraction Reagent is the first commercially available yeast lysis reagent to use a mild detergent lysis procedure to efficiently release functionally active solubilized proteins.

This detergent-based cell lysis buffer eliminates the need to use glass beads or mechanical disruption to break through the thick proteinaceous cell envelope to extract protein Y-PER Reagent is effective for S. cerevisiae and other popular species, making it applicable for use in fusion-tagged protein purification and reporter enzyme assays with these model organisms. The lysis reagent also can be used for genomic and plasmid DNA extraction from yeast. The standard protocol for protein extraction is easy: simply add an appropriate volume of Y-PER Reagent to pelleted yeast cells, incubate at room temperature for approximately 20 minutes and spin down the debris. The resulting supernatant is a concentrated protein solution, surpassing typical yields obtained with traditional glass bead disruption.

Highlights:

  • More protein – extracts more than twice as much protein as glass bead methods

  • Less hassle – eliminates the difficulties associated with traditional glass bead lysis (e.g., clinging static-charged beads, protein/bead clumps and runaway beads)

  • Optmized for yeast – validated for use with Saccharomyces cerevisiae, Schizosaccharomyces pombe and Pichia pastoris

  • Versatile – effective for many different organisms, including Bacillus subtilus and both Gram-positive and Gram-negative bacteria; suitable for use in a diverse range of situations

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 mL
Format Liquid
Product Type Yeast Protein Extraction Reagent
Volume (Metric) 200 mL
Content And Storage Store product at room temperature.
Product Line Y-PER
compliance-icons
Product Identifier
  • Y-PER Yeast Protein Extraction Reagent
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : glycerol, disodium hydrogenorthophosphate

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.