missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yeast Extract, Ultrapure, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
Description
- Format: Off-white powder
- CAS: 8013-01-2
- MDL Number: MFCD00132599
Specifications
Color | Off-white |
Form | Powder |
Grade | Ultrapure |
Product Type | Microbiology Media |
Packaging Type | Plastic Bottle |
Quantity | 1 kg |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'