missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEPperCHIP™ Yellow Fever Virus Proteome Microarray
Yellow Fever Virus Proteome Microarray with 3834 virus specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.
Brand: PEPperCHIP™ SPC.026.001
This item is not returnable.
View return policy
Description
- Microarray Content: Genome polyproteins of yellow fever virus are cleaved into 14 chains: capsid protein C, prM, peptide pr, small envelope protein M, envelope protein E, non-structural protein 1, non-structural protein 2A, non-structural protein 2A-alpha, serine protease subunit NS2B, serine protease NS3, non-structural protein 4A, peptide 2k, non-structural protein 4B, RNA-directed RNA polymerase NS5
- Peptide Length/Overlap: 15 aa/ 14 aa
- Number of Peptides: 3834 printed in duplicate and HA, polio and c-Myc control peptides
Specifications
PEGMA/PMMA | |
1 Slide | |
1 mm | |
Transparent | |
Individual barcode for each microarray | |
High-density Peptide Microarray | |
Non-sterile |
1 array copy per microarray | |
Combinatorial peptide synthesis by amino acid laser printing | |
ISO 9001:2015 | |
RUO | |
Microscopic Glass | |
Antibody validation, epitope mapping, serum screening, biomarker discovery | |
75.4 x 25 mm |