missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Yellow Link

Product Code. 11776925
Click to view available options
Color:
Yellow
Unit Size:
1 pack
This item is not returnable. View return policy

Product Code. 11776925

Brand: Brady™ 292252

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This Yellow Rubber Link enables you to attach a second chain to your posts.

  • This yellow rubber link enables you to attach a second chain to your posts.

Specifications

Product Type Chain Link
Alternate Part Number BRADYLINK RUBBER YELLOW LINK PK 20
Color Yellow
Description Link
For Use With (Application) Traffic Control
Quantity 20 Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.