missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Yellow Zirconium Oxide Beads
Yellow Zirconium Oxide Beads are effective at breaking very tough and hard samples, including organisms within a tough or dense exterior matrix.
Brand: MP Biomedicals™ 116540442
This item is not returnable.
View return policy
Description
- Useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing
- For isolation of RNA and Proteins from bacterial aggregates, biofilms, and whole insects
- Sample Type: Plants, Animal Tissues, Wood, Seeds, Soil, Clay, Bacteria
Specifications
Zirconium Oxide | |
Room temperature | |
FastPrep-24 5G, FastPrep-24, FastPrep-96 | |
Yellow Zirconium Oxide Bead |
Yellow | |
Plants, Animal Tissues, Wood, Seeds, Soil, Clay, Bacteria | |
Extractions of nucleic acids, proteins, metabolites and other small molecules | |
1 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction