missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lauda™ Yellowish Clear Heat Transfer Liquid
Brand: Lauda™ LZB 107
This item is not returnable.
View return policy
Specifications
5 L | |
Bath Filler | |
5 L |
30°C to 200°C | |
Ultra 350 | |
47mm2/sec. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Bath liquid ULTRA 350 (1 can à 5 litres) .
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Dibenzylbenzene, ar-methyl derivitive
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction