missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ YPD Broth
GREENER_CHOICE

Product Code. 12094108
Click to view available options
Quantity:
1 L
Unit Size:
1000mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 12094108

Brand: Gibco™ A1374501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

YPD Broth

Gibco™ YPD Broth is a ready-to-use liquid growth media used for yeast culture. Our filter-sterilized Gibco™ YPD Broth can be used for nearly all research applications that require a liquid yeast culturing step such as protein expression, cloning, and expansion.
  • Gibco™ microbiological media
  • Convenient ready-to-use liquid format
  • 1 L size available

    YPD Broth Recipe
    YPD Broth is a commonly used yeast growth media for maintenance and propagation of Pichia pastoris (P. pastoris) and Saccharomyces cerevisae (S. cerevisae) yeast strains.

    Formulation per One Liter:
    10 g   Yeast extract
    20 g   Bacto peptone
    20 g   Dextrose (glucose)

    Ready-to-Use in Optimized Gibco™ Media Bottles
    Each bottle of Gibco™ YPD Broth is filter-sterilized and ready-to-use in ergonomic Gibco™ media bottles with angled openings for simplified pipetting.

    For research use only. Not intended for human or animal therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage 1 liter YPD Broth, liquid; store at 2 to 8 °C. Protect from light.
    Form Liquid
    Media Type YPD
    Preparation Method Ready-to-Use
    Product Type Microbiology Media
    Sterility Sterile
    Target Organism Class P. pastoris, S. cerevisiae
    Final Product Type Liquid Medium
    Product Line Gibco
    Quantity 1 L
    Shipping Condition Room Temperature
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.