Learn More
YSI™ Probe Service Kits for YSI Dissolved Oxygen Probes
For replenishing oxygen sensor on YSI Dissolved Oxygen Probes
£53.30 - £147.90
Specifications
Probe Type DO
View More Specs
Products 4
Product Code Brand For Use With (Equipment) Quantity Type Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Quantity Type Price Quantity & Availability  
12714576
View Documents
YSI™
066155
Replenishing Oxygen Sensor on YSI™ DO Probe 10/pk. Standard Membrane Kit: 150 membranes This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12724576
View Documents
YSI™
066156
YSI™ 55, 5739, 5750, 5718, 6030 and 6562 DO Probe 10/pk. High Sensitivity Membrane Kit: 150 membranes
£98.30
Each
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
12704576
View Documents
YSI™
098095
Replenishing Oxygen Sensor on YSI™ DO Probe Each High Sensitivity Membrane Kit: 30 membranes, KCl
£62.40
Each
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
12794566
View Documents
YSI™
098094
5718, 5739, 5750, 5300 and Rapid Pulse Probes Each Standard Membrane Kit: 30 membranes, KCl
£53.30
Each
Estimated Shipment: 12-02-2025
to see stock.
Add to basket
Description
Description
  • Membrane Kits include 150 standard, 1mil PTFE membranes or 150 high-sensitivity, 0.5mil PTFE membranes, in 10 packs of 50
  • Membrane/KCI Kits include 30 standard or high-sensitivity membranes, 1 oz. (30mL) dropper bottle with KCI crystals for adding distilled water and a probe contour tool
  • For use with probes 13-299-43 (Model 5739), 13-298-80 (Model 5750), 15-176-50 (Model 5718), 13-298-16 (Model 5300 Biological Oxygen Monitor), and 13-298-56, -57, or -62 (Model 55 Handheld Dissolved Oxygen Meters)
Specifications
Specifications
DO