missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ytterbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Description
Used for phosphors, ceramic capacitors, ferrite devices, pulsed lasers and catalysts. Use in improving the strength and mechanical properties of stainless steel. Used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
UN Number | UN3178 |
Quantity | 50 x 50 mm |
Solubility Information | It reacts slowly with water and is soluble in dilute acids. |
Percent Purity | 99.9% |
Odor | Odorless |
Packaging | Mylar under Argon |
Weight | ≈0.43g/25x25mm |
Form | Foil |
Assay | (REO) |
Chemical Name or Material | Ytterbium foil, 0.1mm (0.004 in.) thick |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'