Learn More
Ytterbium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Description
Yttrium Acetate is widely applied in electronic ceramics, glass, and electronics. Yttrium compounds are used as a catalyst for ethylene polymerization. Yttrium is used in the production of a large variety of synthetic garnets, and Yttria is used to make Yttrium Iron Garnets, which are very effective microwave filters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Crystalline Aggregates |
Quantity | 50 g |
Solubility Information | Insoluble in water,moderately soluble in strong mineral acids. |
Formula Weight | 350.16 (Anhydrous) |
Percent Purity | 99.9% |
Grade | Reacton™ |
Odor | Odorless |
Assay | (REO) |
Chemical Name or Material | Ytterbium(III) acetate hydrate |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W06J032 | Certificate of Analysis | 22/09/2023 | 014576.09,014576.18 |
S20J048 | Certificate of Analysis | 13/07/2023 | 014576,014576.09,014576.18 |
R26H013 | Certificate of Analysis | 12/04/2022 | 014576.09,014576.18 |
A10X010 | Certificate of Analysis | 28/12/2021 | 014576.09,014576.18 |
C19M63 | Certificate of Analysis | 08/12/2021 | 014576.09,014576.18 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.