missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) bromide hydrate, 99.9% (REO)

Product Code. 11332528
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11332528

Brand: Thermo Scientific Alfa Aesar 014575.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15163-03-8

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15163-03-8
MDL Number MFCD00049613
Synonym ytterbium bromide hydrate, acmc-20aj9h, ytterbium 3+ bromide-water 1/3/1

Specifications

Melting Point 300°C
Color White
Physical Form Crystalline
Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 412.77 (Anhydrous)
Percent Purity 99.9%
Sensitivity Very Hygroscopic
Assay (REO)
Chemical Name or Material Ytterbium(III) bromide hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.