missing translation for 'onlineSavingsMsg'
Learn More
Ytterbium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11302528

Ytterbium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11302528
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11302528

Brand: Thermo Scientific Alfa Aesar 014574.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00050146

Specifications

Melting Point (decomposition)
Physical Form Crystalline
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 526.11 (Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Assay (REO)
Chemical Name or Material Ytterbium(III) carbonate hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberG31Z014 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number014574.18,014574.09
Lot NumberF27K30 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014574.18,014574.09
Lot NumberC11Z046 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number014574.18,014574.09
Lot NumberI13S030 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number014574.18,014574.09
Lot NumberI15W032 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number014574.18,014574.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ytterbium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.