missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) chloride, 35% min w/w aq. soln., 99.9% (REO)

Product Code. 11310008
Click to view available options
Quantity:
50 mL
250 mL
1 L
Unit Size:
1 L
250mL
50mL
This item is not returnable. View return policy

Product Code. 11310008

Brand: Thermo Scientific Alfa Aesar 042816.AD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10361-91-8 | YbCl3

Applied with NiCl2 to form a very effective catalyst for the reductive dehalogenation of aryl halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-91-8
Molecular Formula YbCl3
MDL Number MFCD00011288

Specifications

Physical Form Liquid
Quantity 50 mL
Solubility Information Soluble in water.
Formula Weight 279.40 (Anhydrous)
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 35% min w/w aq. soln.
Odor Odorless
Density 1.48 g/mL at 30°C
Assay (REO)
Chemical Name or Material Ytterbium (III) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.