missing translation for 'onlineSavingsMsg'
Learn More
Ytterbium(III) chloride, 35% min w/w aq. soln., 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11330008

Ytterbium(III) chloride, 35% min w/w aq. soln., 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11330008
250 mL
Click to view available options
Quantity:
50 mL
250 mL
1 L
This item is not returnable. View return policy

Product Code. 11330008

Brand: Thermo Scientific Alfa Aesar 042816.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applied with NiCl2 to form a very effective catalyst for the reductive dehalogenation of aryl halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-91-8
Molecular Formula YbCl3
MDL Number MFCD00011288

Specifications

Physical Form Liquid
Quantity 250 mL
Solubility Information Soluble in water.
Formula Weight 279.40 (Anhydrous)
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 35% min w/w aq. soln.
Odor Odorless
Density 1.48 g/mL at 30°C
Assay (REO)
Chemical Name or Material Ytterbium (III) chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberL04K26 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042816.K2,042816.AD,042816.AK
Lot NumberK22J09 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number042816.K2,042816.AD,042816.AK
Lot NumberK22M24 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number042816.K2,042816.AD,042816.AK
Lot NumberA31L16 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number042816.K2,042816.AD,042816.AK

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ytterbium(III) chloride, 35% min w/w aq. soln., 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.