missing translation for 'onlineSavingsMsg'
Learn More
Ytterbium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11366218

Ytterbium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11366218
1 g
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11366218

Brand: Thermo Scientific Alfa Aesar 011189.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ytterbium(III) fluoride is used in the preparation and study of fluorinated glasses. Further, it is employed as a laboratory reagent and in optical glasses and structural ceramics. It finds application in catalysts, electrical components and photo-optical materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-80-0
Molecular Formula F3Yb
Molecular Weight (g/mol) 230.04
MDL Number MFCD00049615
InChI Key XASAPYQVQBKMIN-UHFFFAOYSA-K
Synonym ytterbium 3+ trifluoride, ytterbium 3+ ion trifluoride, acmc-209ceg, ytterbium iii ; fluoride, fluoride, fluoride, yb, fluoride, ytterbium iii fluoride, anhydrous trace rare earth metals basis 10gShow MoreShow Less
PubChem CID 21694467
SMILES [F-].[F-].[F-].[Yb+3]

Specifications

Melting Point 1157°C
Physical Form Lump/Powder
Quantity 1 g
UN Number UN3288
Solubility Information Slightly soluble in strong mineral acids. Insoluble in water.
IUPAC Name ytterbium(3+) trifluoride
Formula Weight 230.04
Percent Purity 99.99%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 8.2 g/mL
Assay (REO)
Chemical Name or Material Ytterbium(III) fluoride, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT03D029 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog Number011189.03,011189.09,011189.18
Lot Number10250644 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number011189.03,011189.09,011189.18
Lot NumberT03D029 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011189.03,011189.09,011189.18
Lot NumberG22W039 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011189.03,011189.09,011189.18
Lot NumberJ22W018 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011189.03,011189.09,011189.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ytterbium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.