missing translation for 'onlineSavingsMsg'
Learn More
Ytterbium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11366228

Ytterbium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11366228
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11366228

Brand: Thermo Scientific Alfa Aesar 011196.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ytterbium(III) nitrate hydrate, is used as a precursor of nanoscale coatings of carbon composites. Source of yttrium used to make yttrium-based surfactant mesophases which are promising as adsorbing agents as well as for optically functional species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00149883

Specifications

Color White
Physical Form Crystalline Aggregates
Quantity 10 g
UN Number UN1477
Merck Index 14,10105
Solubility Information Soluble in water.
Formula Weight 359.05 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Ytterbium(III) nitrate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN14L019 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog Number011196.09,011196.18
Lot NumberZ03H068 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011196.09,011196.18
Lot NumberF06T019 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011196.09,011196.18
Lot NumberB11X013 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011196.09,011196.18
Lot NumberI28J12 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011196.09,011196.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ytterbium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.