missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) triflate, 99.9%, (trace metal basis), anhydrous

Product Code. p-4433124 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11914381

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54761-04-5 | C3F9O9S3Yb | 620.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54761-04-5
Molecular Formula C3F9O9S3Yb
Molecular Weight (g/mol) 620.25
InChI Key AHZJKOKFZJYCLG-UHFFFAOYSA-K
Synonym ytterbium iii trifluoromethanesulfonate, ytterbium iii triflate, ytterbium triflate, unii-4n70osw24g, ytterbium trifluoromethanesulfonate, yb otf 3, ytterbium 3+ tritriflate, trifluoromethanesulfonate; ytterbium 3+, ytterbium 3+ triflate, ytterbium trifluoromethanesulphonate
PubChem CID 2733225
IUPAC Name trifluoromethanesulfonate;ytterbium(3+)
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Yb+3]

Specifications

Quantity 1 g
Assay Percent Range 26.8 to 29% (Yb) (Complexometry)
Linear Formula (CF3SO3)3Yb
Merck Index 15,10304
Solubility Information Solubility in water: soluble
Formula Weight 620.25
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ytterbium(III) triflate
compliance-icons
Product Identifier
  • Ytterbium(III) triflate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.