missing translation for 'onlineSavingsMsg'
Learn More
Ytterbium(III) triflate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10045913

Ytterbium(III) triflate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10045913
25 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10045913

Brand: Thermo Scientific Acros 304170250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 252976-51-5
Molecular Formula C3F9O9S3Yb
Molecular Weight (g/mol) 620.23
MDL Number MFCD06200261
InChI Key AHZJKOKFZJYCLG-UHFFFAOYSA-K
Synonym ytterbium iii trifluoromethanesulfonate hydrate, ytterbium 3+ hydrate tritriflate, ytterbium iii triflate hydrate, ytterbium triflate hydrate, acmc-1aqes, yb otf 3.h2o, ytterbium triflate monohydrate, ytterbium trifluoromethanesulfonate hydrate, ytterbium trifluoromethanesulfonate, hydrateShow MoreShow Less
PubChem CID 22836430
IUPAC Name ytterbium(3+) tritrifluoromethanesulfonate
SMILES [Yb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point 118.0°C to 122.0°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99.90%
Linear Formula (CF3SO3)3Yb·xH2O
Solubility Information Solubility in water: soluble
Formula Weight 620.23
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ytterbium(III) triflate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466770 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog Number304170050,304170250
Lot NumberA0433651 Certificate TypeCertificate of Analysis Date26/02/2024 Catalog Number304170050,304170250,304170010
Lot NumberA0450821 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number304170050,304170250
Lot NumberA0384232 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number304170050,304170250,304170010
Lot NumberA0409257 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number304170050,304170250,304170010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ytterbium(III) triflate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.