missing translation for 'onlineSavingsMsg'
Learn More
Ytterbium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 15400967

Ytterbium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 15400967
10 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 15400967

Brand: Thermo Scientific Alfa Aesar 000408.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

packaged under argon in glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-64-4
Molecular Formula Yb
Molecular Weight (g/mol) 173.05
MDL Number MFCD00011286
InChI Key NAWDYIZEMPQZHO-UHFFFAOYSA-N
Synonym unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
PubChem CID 23992
ChEBI CHEBI:33381
IUPAC Name ytterbium
SMILES [Yb]

Specifications

Physical Form Powder
Quantity 10 g
Assay Percent Range 99.9% (REO)
Formula Weight 173.04
Sensitivity Air and moisture sensitive
Density 6.97
Chemical Name or Material Ytterbium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW05J033 Certificate TypeCertificate of Analysis Date14/09/2023 Catalog Number000408.04,000408.09
Lot NumberM21I029 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number000408.04,000408.09
Lot NumberB16S011 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000408.04,000408.09
Lot NumberC16R028 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000408.04,000408.09
Lot NumberK07R059 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000408.04,000408.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ytterbium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.