missing translation for 'onlineSavingsMsg'
Learn More
Yttrium aluminum oxide, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11353298

Yttrium aluminum oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11353298
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11353298

Brand: Thermo Scientific Alfa Aesar 036241.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Can be used as a sputtering target to prepare bubble memory devices

Can be used as a sputtering target to prepare bubble memory devices.Yttrium aluminum oxide finds application in ceramic, glass and optics due to its thermal stability and insolubility. It is used in the preparation of sputtering bonded targets in chemical processes viz. chemical vapor deposition, atomic thermal and electron beam evaporation. It is also used in solar, water treatment and fuel cell. Further, it is used as coloring and adhesive agents in enamels manufacture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12005-21-9
MDL Number MFCD00213958
Synonym Yttrium aluminum garnet; YAG

Specifications

Melting Point 1760°C
Physical Form Fused Lump
Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 593.62
Percent Purity 99.99%
Refractive Index 1.833
Density 4.65 g/mL
Assay (metals basis)
Chemical Name or Material Yttrium aluminum oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium aluminum oxide, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.