missing translation for 'onlineSavingsMsg'
Learn More

Yttrium foil, 0.127mm (0.005in) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11345699
50 x 50 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]

Specifications

Quantity 50 x 50 mm
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.36g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Yttrium foil, 0.127mm (0.005 in.) thick

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium foil, 0.127mm (0.005in) thick, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.