missing translation for 'onlineSavingsMsg'
Learn More
Yttrium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11331479

Yttrium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11331479
25 x 25 mm
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy

Product Code. 11331479

Brand: Thermo Scientific Alfa Aesar 013960.FF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx. 0.7g/25 x 25mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]

Specifications

Color Gray
Appearance Gray foil
Assay Percent Range 99.90%
Quantity 25 x 25 mm
Odor Odorless
Form Foil
Chemical Name or Material Yttrium, Rare Earth Oxides Content

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.