missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium(III) 2-ethylhexanoate, 99.8% (metals basis), Thermo Scientific Chemicals
£35.30 - £442.00
Chemical Identifiers
CAS | 103470-68-4 |
---|---|
Molecular Formula | C24H45O6Y |
Molecular Weight (g/mol) | 518.524 |
MDL Number | MFCD00070441 |
InChI Key | AGOMHFKGCMKLDA-UHFFFAOYSA-K |
Synonym | yttrium iii 2-ethylhexanoate, yttrium 2-ethylhexanoate, hexanoic acid,2-ethyl-, yttrium 3+ salt 3:1, yttrium iii 2-ethylhexanoate w/v in n-hexane, acmc-1c5ry, yttrium tri 2-ethylhexanoate, yttrium tris 2-ethylhexanoate, tris 2-ethylhexanoic acid yttrium salt, yttrium 3+ tris 2-ethylhexanoate, yttrium iii 2-ethylhexanoate y |
PubChem CID | 15607674 |
IUPAC Name | 2-ethylhexanoate;yttrium(3+) |
SMILES | CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Y+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11388088
|
Thermo Scientific Alfa Aesar
036661.06 |
5 g |
£35.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11398088
|
Thermo Scientific Alfa Aesar
036661.14 |
25 g |
£136.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11378088
|
Thermo Scientific Alfa Aesar
036661.22 |
100 g |
£442.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Yttrium(III) 2-ethylhexanoate has been used for research in nanotechnology based applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
103470-68-4 | |
518.524 | |
AGOMHFKGCMKLDA-UHFFFAOYSA-K | |
15607674 | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Y+3] |
C24H45O6Y | |
MFCD00070441 | |
yttrium iii 2-ethylhexanoate, yttrium 2-ethylhexanoate, hexanoic acid,2-ethyl-, yttrium 3+ salt 3:1, yttrium iii 2-ethylhexanoate w/v in n-hexane, acmc-1c5ry, yttrium tri 2-ethylhexanoate, yttrium tris 2-ethylhexanoate, tris 2-ethylhexanoic acid yttrium salt, yttrium 3+ tris 2-ethylhexanoate, yttrium iii 2-ethylhexanoate y | |
2-ethylhexanoate;yttrium(3+) |
Specifications
103470-68-4 | |
MFCD00070441 | |
Insoluble in water. | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Y+3] | |
518.524 | |
518.16 | |
Powder | |
Yttrium(III) 2-ethylhexanoate |
C24H45O6Y | |
yttrium iii 2-ethylhexanoate, yttrium 2-ethylhexanoate, hexanoic acid,2-ethyl-, yttrium 3+ salt 3:1, yttrium iii 2-ethylhexanoate w/v in n-hexane, acmc-1c5ry, yttrium tri 2-ethylhexanoate, yttrium tris 2-ethylhexanoate, tris 2-ethylhexanoic acid yttrium salt, yttrium 3+ tris 2-ethylhexanoate, yttrium iii 2-ethylhexanoate y | |
AGOMHFKGCMKLDA-UHFFFAOYSA-K | |
2-ethylhexanoate;yttrium(3+) | |
15607674 | |
99.8% | |
(metals basis) |
Safety and Handling
GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
P201-P202-P281-P308+P313-P501c
H361d
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only