missing translation for 'onlineSavingsMsg'
Learn More
Yttrium(III) acetate tetrahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11356208

Yttrium(III) acetate tetrahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11356208
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11356208

Brand: Thermo Scientific Alfa Aesar 011186.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Yttrium(III) acetate tetrahydrate is used as laboratory reagents, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85949-60-6
Molecular Formula C6H17O10Y
Molecular Weight (g/mol) 338.10
MDL Number MFCD00013048
InChI Key AIQRTHPXPDTMBQ-UHFFFAOYSA-K
IUPAC Name yttrium(3+) triacetate tetrahydrate
SMILES O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Melting Point (decomposition)
Physical Form Crystalline Aggregates
Quantity 100 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in Hot water.
Formula Weight 338.10 (266.04 Anhydrous)
Density 1.5 g/mL
Chemical Name or Material Yttrium(III) acetate tetrahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium(III) acetate tetrahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.