missing translation for 'onlineSavingsMsg'
Learn More
Yttrium(III) chloride hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10216640

Yttrium(III) chloride hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10216640
250 g, Glass bottle
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10216640

Brand: Thermo Scientific Acros 199182500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-94-2
Molecular Formula Cl36H2O
Molecular Weight (g/mol) 303.35
MDL Number MFCD00149941

Specifications

Melting Point 100.0°C
Color White
Physical Form Adhering Crystalline Powder or Crystals
Quantity 250 g
Assay Percent Range 99.90%
Linear Formula YCl3·6H2O
Merck Index 14, 10107
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols
Formula Weight 303.35
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Yttrium(III) chloride hexahydrate, 99.9%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468197 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number199182500,199180500
Lot NumberA0467020 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number199182500,199180500
Lot NumberA0463681 Certificate TypeCertificate of Analysis Date20/06/2024 Catalog Number199182500,199180500
Lot NumberA0458333 Certificate TypeCertificate of Analysis Date30/11/2023 Catalog Number199182500,199180500
Lot NumberA0458039 Certificate TypeCertificate of Analysis Date21/11/2023 Catalog Number199182500,199180500
compliance-icons
Product Identifier
  • Yttrium(III) chloride hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium(III) chloride hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.