Learn More
Yttrium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Description
Yttrium(III) chloride hydrate acts as a source to prepare highly active and enantioselective yttrium precatalysts for asymmetric hydroamination of gem-disubstituted aminoalkenes. It is used as a precursor for the synthesis of yttrium oxide nanocrystallites. It employs in the crystal structure determination of macromolecules such as proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Wet Crystalline Aggregates |
Quantity | 500 g |
Merck Index | 14,10107 |
Solubility Information | Soluble in water. |
IUPAC Name | yttrium(3+);trichloride;hydrate |
Formula Weight | 195.26 (Anhydrous) |
Percent Purity | 99.9% |
Grade | Reacton™ |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y14K015 | Certificate of Analysis | 19/11/2024 | 011184.22,011184.14,011184.36 |
B02S039 | Certificate of Analysis | 12/11/2021 | 011184.22,011184.14,011184.36 |
G01W013 | Certificate of Analysis | 12/11/2021 | 011184.22,011184.14,011184.36 |
L17P30 | Certificate of Analysis | 12/11/2021 | 011184.22,011184.14,011184.36 |
B01Q03 | Certificate of Analysis | 12/11/2021 | 011184.22,011184.14,011184.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.