Learn More
Yttrium(III) iodide, ultra dry, 99.9% (REO)
Description
Yttrium(III) iodide, is used as a starting material for YBCO superconducting materials in a lower temperature preparation. Applied in the solid-state synthesis of a new compound, Y21I18C14B7, which is the first example of a rare earth metal halide with interstitial halogen atoms with misfit characteristics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
| Melting Point | 1,004°C |
| Physical Form | Chunks and/or Powder |
| Quantity | 5 g |
| Solubility Information | Soluble in water. |
| Formula Weight | 469.62 |
| Percent Purity | 99.9% |
| Packaging | Ampule |
| Sensitivity | Hygroscopic |
| Assay | (REO) |
| Chemical Name or Material | Yttrium(III) iodide, Ultra dry |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.