missing translation for 'onlineSavingsMsg'
Learn More
Yttrium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11327637

Yttrium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11327637
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
5 x 100 g
This item is not returnable. View return policy

Product Code. 11327637

Brand: Thermo Scientific Alfa Aesar 012898.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as a reagent, in optics, ceramics, glass and electronics. It is used in superconducting materials. It has been reported as a reusable catalyst in Bignelli reaction for the synthesis of heterocycles such as pyrimidin-2-ones. It is a powerful catalyst for the synthesis of some supramolecules such as calix[4]resorcinarenes, 1,8-dioxooctahydroxanthenes, 2-amino-4H-chromenes, and several other organic condensation products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13494-98-9
Molecular Formula H12N3O15Y
Molecular Weight (g/mol) 383.008
MDL Number MFCD00149940
InChI Key QBAZWXKSCUESGU-UHFFFAOYSA-N
Synonym yttrium nitrate hexahydrate, yttrium iii nitrate hexahydrate, yttriumnitrat german, yttrium iii nitrate hexahydrate 1:3:6, yttrium trinitrate hexahydrate, nitric acid, yttrium 3+ salt, hexahydrate, nitric acid,yttrium 3+ salt, hexahydrate 8ci,9ci, yttriumnitrat, yttrium iii nitrate hexahydrate reo, ytterium nitrateShow MoreShow Less
PubChem CID 166833
IUPAC Name yttrium(3+);trinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3]

Specifications

Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water.
Formula Weight 383.01 (274.92 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Yttrium(III) nitrate hexahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU17C040 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number012898.14,012898.22,012898.06,012898.59
Lot NumberR24K025 Certificate TypeCertificate of Analysis Date24/06/2024 Catalog Number012898.14,012898.22,012898.06,012898.59
Lot NumberR25K025 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number012898.14,012898.22,012898.06,012898.59
Lot NumberZ14J014 Certificate TypeCertificate of Analysis Date11/01/2024 Catalog Number012898.14,012898.22,012898.06,012898.59
Lot NumberN08J013 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number012898.14,012898.22,012898.06,012898.59

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.