Learn More
Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
£38.25 - £361.80
Chemical Identifiers
CAS | 13494-98-9 |
---|---|
Molecular Formula | N3O9Y·6H2O |
Molecular Weight (g/mol) | 383.01 |
MDL Number | MFCD00149940 |
InChI Key | QBAZWXKSCUESGU-UHFFFAOYSA-N |
Synonym | yttrium nitrate hexahydrate, yttrium iii nitrate hexahydrate, yttriumnitrat german, yttrium iii nitrate hexahydrate 1:3:6, yttrium trinitrate hexahydrate, nitric acid, yttrium 3+ salt, hexahydrate, nitric acid,yttrium 3+ salt, hexahydrate 8ci,9ci, yttriumnitrat, yttrium iii nitrate hexahydrate reo, ytterium nitrateShow More |
PubChem CID | 166833 |
IUPAC Name | yttrium(3+);trinitrate;hexahydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10338210
|
Thermo Scientific Acros
218260250 |
25 g | Glass bottle |
£38.25
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10308260
|
Thermo Scientific Acros
218261000 |
100 g | Glass bottle |
£106.61
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10626214
|
Thermo Scientific
218265000 |
500 g | Glass bottle |
£361.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
13494-98-9 | |
383.01 | |
QBAZWXKSCUESGU-UHFFFAOYSA-N | |
166833 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3] |
N3O9Y·6H2O | |
MFCD00149940 | |
yttrium nitrate hexahydrate, yttrium iii nitrate hexahydrate, yttriumnitrat german, yttrium iii nitrate hexahydrate 1:3:6, yttrium trinitrate hexahydrate, nitric acid, yttrium 3+ salt, hexahydrate, nitric acid,yttrium 3+ salt, hexahydrate 8ci,9ci, yttriumnitrat, yttrium iii nitrate hexahydrate reo, ytterium nitrateShow More | |
yttrium(3+);trinitrate;hexahydrate |
Specifications
13494-98-9 | |
Adhering Crystalline Powder or Crystals | |
N3O9Y·6H2O | |
MFCD00149940 | |
yttrium nitrate hexahydrate, yttrium iii nitrate hexahydrate, yttriumnitrat german, yttrium iii nitrate hexahydrate 1:3:6, yttrium trinitrate hexahydrate, nitric acid, yttrium 3+ salt, hexahydrate, nitric acid,yttrium 3+ salt, hexahydrate 8ci,9ci, yttriumnitrat, yttrium iii nitrate hexahydrate reo, ytterium nitrate | |
QBAZWXKSCUESGU-UHFFFAOYSA-N | |
yttrium(3+);trinitrate;hexahydrate | |
166833 | |
99.9% | |
Yttrium(III) nitrate hexahydrate, 99.9% |
White | |
99.9% | |
Y(NO3)3·6H2O | |
14,10107 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3] | |
383.01 | |
383.01 | |
Trace Metal Basis |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medic
GHS Signal Word: Warning
missing translation for 'rtecsNumber' : ZG3750000
- Yttrium(III) nitrate hexahydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.