missing translation for 'onlineSavingsMsg'
Learn More
Yttrium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11396208

Yttrium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11396208
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
5 x 100 g
This item is not returnable. View return policy

Product Code. 11396208

Brand: Thermo Scientific Alfa Aesar 011187.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It acts as a precursor for the preparation of yttrium oxide nano particles and for nanoscale coatings of carbon composites. It is an effective yttrium source for the preparation of yttrium-based surfactant mesophases which are used as adsorbing agents and in optically functional materials. It is used in optics, ceramics, glass, electronics, and superconducting materials. It has been reported as a reusable catalyst in Bignelli reaction for the synthesis of heterocycles such as pyrimidin-2-ones. It is a powerful catalyst for the synthesis of some supramolecules such as calix[4]resorcinarenes, 1,8-dioxooctahydroxanthenes, 2-amino-4H-chromenes, and several other organic condensation products. Silica-supported yttrium nitrate hydrate acts as a heterogeneous catalyst for the preparation of 2-Amino-4H-Chromenes in water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140191-40-8
Molecular Formula N3O9Y
Molecular Weight (g/mol) 274.92
MDL Number MFCD00011469
InChI Key BXJPTTGFESFXJU-UHFFFAOYSA-N
Synonym yttrium iii nitrate hydrate, yttrium nitrate hydrate, 3no3.y.h2o, nitric acid, yttrium iii salt, nitric acid, yttrium 3++ salt, yttrium 3+ hydrate trinitrate, yttrium iii nitrate hydrate, reactonShow MoreShow Less
PubChem CID 23169389
SMILES [Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 100 g
UN Number UN1477
Merck Index 14,10107
Solubility Information Soluble in water.
IUPAC Name yttrium(3+);trinitrate;hydrate
Formula Weight 274.92 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Yttrium(III) nitrate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS05K067 Certificate TypeCertificate of Analysis Date25/06/2024 Catalog Number011187.22,011187.06,011187.14,011187.59
Lot NumberY09G053 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number011187.22,011187.06,011187.14,011187.59
Lot NumberX11J010 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number011187.22,011187.06,011187.14,011187.59
Lot Number61200296 Certificate TypeCertificate of Analysis Date23/11/2022 Catalog Number011187.22,011187.06,011187.14,011187.59
Lot Number61200296 Certificate TypeCertificate of Analysis Date22/11/2022 Catalog Number011187.22,011187.06,011187.14,011187.59

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.