Learn More
Yttrium(III) oxalate nonahydrate, 99.9% (REO), Thermo Scientific Chemicals
Description
Yttrium oxalate is applied in ceramics, glass, and electronics. It is an important material for tri-bands rare Earth phosphors and yttrium-iron-garnets, which are very effective microwave filters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | (decomposition) |
Color | White |
Physical Form | Crystalline |
Quantity | 50 g |
Linear Formula | Y2(C2O4)3·9H2O |
UN Number | UN3288 |
Solubility Information | Insoluble in water,moderately soluble in strong mineral acids |
Formula Weight | 604.01 (441.87 Anhydrous) |
Percent Purity | 99.9% |
Assay | (REO) |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y15K015 | Certificate of Analysis | 06/12/2024 | 014562.18 |
U09K058 | Certificate of Analysis | 14/08/2024 | 014562.18 |
P05K017 | Certificate of Analysis | 12/03/2024 | 014562.18 |
N05K002 | Certificate of Analysis | 16/02/2024 | 014562.18 |
U09J054 | Certificate of Analysis | 24/10/2023 | 014562.18 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.