missing translation for 'onlineSavingsMsg'
Learn More
Yttrium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 15720829

Yttrium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 15720829
250 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 15720829

Brand: Thermo Scientific Acros 194612500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Melting Point 2410.0°C
Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 99.99% min. (Y2O3) TREO base
Linear Formula Y2O3
Merck Index 14, 10107
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dilute acids
Formula Weight 225.81
Percent Purity 99.99%
Grade Trace Metal Basis
Loss on Ignition 1.0 % max. (1000°C) (material base)
Packaging Glass bottle
Chemical Name or Material Yttrium(III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467134 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog Number194610500,194612500,194610100
Lot NumberA0457025 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog Number194610500,194612500,194610100
Lot NumberA0404933 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number194610500,194612500,194610100
Lot NumberA0377157 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number194610500,194612500,194610100
Lot NumberA0417054 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number194610500,194612500,194610100

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium(III) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.