missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) oxide, 99.999%, (trace metal basis), Thermo Scientific Chemicals

£16.78 - £144.72

Chemical Identifiers

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10174210
View Documents Promotion Details
Thermo Scientific Acros
194480020
2 g Glass bottle
£16.78
2g
Please to purchase this item. Need a web account? Register with us today!
10328430
View Documents Promotion Details
Thermo Scientific Acros
194480100
10 g Glass bottle
£31.56
10g
Please to purchase this item. Need a web account? Register with us today!
10301054
View Documents Promotion Details
Thermo Scientific Acros
194480500
50 g Glass bottle
£144.72
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Specifications

Chemical Identifiers

1314-36-9
225.81
RUDFQVOCFDJEEF-UHFFFAOYSA-N
[O--].[O--].[O--].[Y+3].[Y+3]
O3Y2
MFCD00011473
diyttrium(3+) trioxidandiide

Specifications

2410.0°C
White
99.999% (Y2O3) On TREO base
Y2O3
14, 10107
RUDFQVOCFDJEEF-UHFFFAOYSA-N
diyttrium(3+) trioxidandiide
225.81
Trace Metal Basis
1314-36-9
Powder
O3Y2
MFCD00011473
Solubility in water: insoluble. Other solubilities: soluble in dilute acids
[O--].[O--].[O--].[Y+3].[Y+3]
225.81
99.999%
Yttrium(III) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 215-233-5

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only