missing translation for 'onlineSavingsMsg'
Learn More
Yttrium(III) oxide pellets, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11348578

Yttrium(III) oxide pellets, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11348578
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11348578

Brand: Thermo Scientific Alfa Aesar 040648.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sintered pellets suitable for vacuum deposition

Sintered pellets suitable for vacuum depositionYttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is a precursor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Melting Point 2410°C
Quantity 100 g
Merck Index 14,10107
Solubility Information Insoluble in water. Soluble in alcohol and dilute acids.
Formula Weight 225.81
Percent Purity 99.9%
Odor Odorless
Sensitivity Air Sensitive
Density 5.01 g/mL
Assay (REO)
Chemical Name or Material Yttrium(III) oxide pellets, approx. 10mm dia. x 5mm thick
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC18M41 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040648.22
Lot NumberC28R046 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040648.22
Lot NumberR30A022 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040648.22
Lot NumberD28R004 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040648.22
Lot NumberB11N16 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040648.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium(III) oxide pellets, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.