missing translation for 'onlineSavingsMsg'
Learn More
Yttrium(III) oxide, REacton™, 99.9999% (REO), Thermo Scientific Chemicals Product Code.: 11320108

Yttrium(III) oxide, REacton™, 99.9999% (REO), Thermo Scientific Chemicals

Product Code. 11320108
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11320108

Brand: Thermo Scientific Alfa Aesar 042864.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Melting Point 2410°C
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.9999% (REO)
Formula Weight 225.81
Density 5.01
Chemical Name or Material Yttrium(III) oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP11L011 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog Number042864.09,042864.18,042864.30
Lot NumberY14K012 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number042864.09,042864.18,042864.30
Lot NumberY01F034 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number042864.09,042864.18,042864.30
Lot NumberG05M16 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number042864.09,042864.18,042864.30
Lot NumberB08R015 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number042864.09,042864.18,042864.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium(III) oxide, REacton™, 99.9999% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.