missing translation for 'onlineSavingsMsg'
Learn More
Yttrium(III) sulfide, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11450231

Yttrium(III) sulfide, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11450231
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11450231

Brand: Thermo Scientific Alfa Aesar 039565.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12039-19-9
Molecular Formula S3Y2
Molecular Weight (g/mol) 273.99
MDL Number MFCD00058806
InChI Key BPMRLDMEAPSVQN-UHFFFAOYSA-N
Synonym yttrium sulfide, diyttrium trisulphide, yttrium iii sulfide, yttrium sulfide y2s3, yttrium 3+ trisulfide, diyttrium 3+ ion trisulfanediide, yttrium iii sulfide trace rare earth metals basis 1gShow MoreShow Less
PubChem CID 166021
SMILES [S--].[S--].[S--].[Y+3].[Y+3]

Specifications

Physical Form Powder
Quantity 25 g
UN Number UN3134
Solubility Information Soluble in water.
IUPAC Name diyttrium(3+) trisulfanediide
Formula Weight 273.93
Percent Purity 99.9%
Odor Like rotten eggs
Assay (REO)
Chemical Name or Material Yttrium(III) sulfide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW11D068 Certificate TypeCertificate of Analysis Date18/01/2025 Catalog Number039565.06,039565.03,039565.14
Lot NumberX16B020 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number039565.06,039565.03,039565.14
Lot NumberK21L22 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039565.06,039565.03,039565.14
Lot NumberW24H040 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039565.06,039565.03,039565.14
Lot NumberK09P11 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039565.06,039565.03,039565.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium(III) sulfide, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.