missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) trifluoroacetate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11485380
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

A fluorinated yttrium compound for proteomics research. Used in the manufacture of ceramics, glass, laser materials, phosphor. non-ferrous alloy iron and steel, and the additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304851-95-4
Molecular Formula C6F9O6Y
Molecular Weight (g/mol) 427.95
MDL Number MFCD05663780 MFCD00192577
InChI Key NPATVYPXYMSPKD-UHFFFAOYSA-K
Synonym yttrium trifluoroacetate
PubChem CID 22836421
IUPAC Name 2,2,2-trifluoroacetic acid;yttrium;hydrate
SMILES [Y+3].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F

Specifications

Physical Form Crystalline
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 427.95 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Yttrium (III) trifluoroacetate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'