missing translation for 'onlineSavingsMsg'
Learn More
Yttrium ingot, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11316547

Yttrium ingot, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11316547
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11316547

Brand: Thermo Scientific Alfa Aesar 000399.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in air stable poly bottle
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]

Specifications

Quantity 10 g
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Contains less than 1% tantalum
Odor Odorless
Packaging Poly bottle
Form Ingot
Assay (REO)
Chemical Name or Material Yttrium ingot

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX13G095 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number000399.09,000399.18
Lot NumberR12I037 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog Number000399.09,000399.18
Lot NumberR20F038 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000399.09,000399.18
Lot NumberD13S033 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000399.09,000399.18
Lot NumberJ19Q048 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000399.09,000399.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium ingot, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.